Learn More
Medchemexpress LLC (2S)-SB02024 | 2126737-29-7 | 345.36 g/mol | C16H22F3N3O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY122891A5MG
Encompass_Preferred
(2S)-SB02024 is the S-enantiomer of SB02024, a small-molecule inhibitor of Vps34 (class III PI3K) provided for laboratory research. The compound has molecular formula C16H22F3N3O2 and molecular weight 345.36 g/mol, and is supplied as a solid in small milligram quantities for in vitro studies. It is intended for research use only and not for clinical applications.
  • Selective Vps34 inhibitor for research use.
  • Provided as a small-quantity solid suitable for biochemical and cellular assays.
  • Characterized by molecular formula and molecular weight for experimental planning.
  • Documentation available including data sheet and handling instructions.
  • Not intended for human or clinical use.
Catalog No. 50-002-63882