Learn More
Medchemexpress LLC (2s)-sb02024 (sb02877), s-enantiomer of sb02024 | 2126737-29-7 | 98.1% | 345.36 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY122891A1MG
Encompass_Preferred
(2S)-SB02024 is the S-enantiomer of the compound SB02024 and functions as an inhibitor of Vps34. Its inhibition of Vps34 has been shown to reprogram cold tumors into hot inflamed tumors and improve anti-PD-1/PD-L1 immunotherapy, suggesting its potential in cancer treatment research.
  • Functions as a Vps34 inhibitor.
  • Reprograms cold tumors into hot inflamed tumors.
  • Enhances anti-PD-1/PD-L1 immunotherapy.
Catalog No. 50-004-36786