Learn More
Medchemexpress LLC (2S)-Selvigaltin | 1978336-95-6 | 99.5% | 533.32 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY147041A50MG
Encompass_Preferred
(2S)-Selvigaltin is the S-enantiomer of Selvigaltin. It is an orally active galectin-3 (Gal-3) inhibitor. This product is for research use only.
  • Inhibits galectin-3.
  • Solid, white to off-white color.
  • Soluble in DMSO at 25 mg/mL (46.88 mM), with ultrasonic, warming, and heating to 60°C.
  • Store at -20°C, protected from light, under nitrogen. In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light, under nitrogen.
Catalog No. 50-003-60424