Learn More
Medchemexpress LLC (2S)-Selvigaltin | 99.5% | 533.32 | 1 MG

Supplier: Medchemexpress LLC HY147041A1MG
(2S)-Selvigaltin is the S-enantiomer of Selvigaltin. It is an orally active galectin-3 (Gal-3) inhibitor, intended for research use only.
- S-enantiomer of Selvigaltin
- Orally active galectin-3 (Gal-3) inhibitor
- Molecular weight of 533.32
- Appears as a white to off-white solid
- Soluble in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.