Learn More
Cayman Chemical 2SOctylahydroxyglutarate
SDP
Supplier:  Cayman Chemical 19695100 UG
Encompass_Preferred
An internal standard for the quantification of (2S)-octyl-α-hydroxyglutarate by GC- or LC-MS
Catalog No. 50-298-2480