Learn More
U.S. Pharmacopeia 2-tert-Butyl-4-hydroxyanisole, 88-32-4, MFCD00057667, 200mg
Supplier:  U.S. Pharmacopeia 1083008
Encompass
Molecular Formula C11H16O2, Molecular Weight 180.245, Boiling Point 507.2 °F (264 °C), Flash point 312.80 °F (156.00 °C), Synonyms: 3-tert-Butyl-4-methoxyphenol, 2-BHA
Catalog No. NC9782960