Learn More
Cayman Chemical 2trans Eicosenoic Acid
SDP
Supplier:  Cayman Chemical 1000762250 MG
Encompass_Preferred
An α,β-unsaturated fatty acid; by-product of the synthesis of Δ2-cis eicosenoic acid
Catalog No. 50-288-9278