Learn More
Alkali Scientific 2XYT Broth, Capsules, 496 Grams of Powder in 32 Total Capsules per Bottle
SDP
Supplier:  Alkali Scientific XYT002CP
Encompass_Preferred
2XYT Broth in 496g of powdered capsules (32 capsules per bottle) is designed to support high-density microbial cultures, ensuring rapid and efficient growth. Each pre-measured capsule makes it easy to prepare large volumes of culture medium for microbial research, cell culture, or industrial fermentation processes. The broth's balanced nutrient profile supports a wide range of organisms, including bacteria and fungi. It is commonly used in microbiological studies that require high yields and rapid growth rates. This packaging provides a convenient and cost-effective solution for laboratories needing consistent culture media with minimal preparation time.
Catalog No. 50-006-60594