Learn More
Chemglass Life Sciences Chemglass Cryogenic Vial, Self-Standing, External Thread, Sterile, 3.0 mL (CGN-4762-004)
SDP
Supplier:  Chemglass Life Sciences CGN4762004
Encompass_Preferred
Chemglass 3.0 mL cryogenic vial with an external thread closure. Supplied sterile and racked in nest format for organized cryogenic storage and long-term low-temperature sample preservation.
  • Round bottom and self-standing design.
  • External thread closure.
  • Sterile; sterilized by E-beam.
  • Racked in 9 x 9 format, 8 racks per case (648 vials per case).
  • Autoclavable.
  • Clear graduations and white marking area.
  • Non-pyrogenic; DNase/RNase free.
  • Solid one-piece construction prevents barcode loss at low temperatures.
  • Recommended for storage in the gas phase of a nitrogen tank.
Catalog No. 50-273-3391