Learn More
Medchemexpress LLC (3-([1,1'-Biphenyl]-2-ylethynyl)-1H-indazol-5-yl)(2,6-diazaspiro[3.5]nonan-2-yl)methanone | 3086536-23-1 | 99.5% | 446.54 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16256550MG
Encompass_Preferred
CDD-2807 is an inhibitor for serine/threonine kinase 33 (STK33) with an IC50 of 9.2 nM. It demonstrates no significant toxicity in mice and can penetrate the blood-testis barrier without accumulating in the brain. CDD-2807 induces a reversible contraceptive effect and has potential for the development of male contraception.
  • Potent inhibitor of serine/threonine kinase 33 (STK33) with an IC50 of 9.2 nM.
  • Can cross the blood-testis barrier.
  • Exhibits no significant toxicity in mice.
  • Does not accumulate in the brain.
  • Induces a reversible contraceptive effect, suggesting potential for male contraceptive development.
  • Intended for research use only.
Catalog No. 50-003-92119