Learn More
Medchemexpress LLC 3-[1-(3-aminopropyl)-1H-indol-3-yl]-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione | 137592-43-9 | 99.0% | 384.43 | C23H20N4O2 | 5 MG

Supplier: Medchemexpress LLC HY1173935MG
Bisindolylmaleimide III is a research-grade small-molecule inhibitor of protein kinase C (PKC) used to study PKCα-dependent signaling in biochemical and cell-based assays. It is supplied as a solid powder and commonly packaged in low-mass vials for experimental use.
- Potent, selective inhibitor of protein kinase C, notably PKCα.
- Molecular weight 384.43 g·mol⁻¹.
- High purity (≈99%).
- Soluble in DMSO (25 mg/mL) with ultrasonic assistance.
- Powder stable at -20°C for extended storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.