Learn More
Medchemexpress LLC 3-[1-(5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-2-carbonyl)piperidin-4-yl]-1H-benzimidazol-2-one | 924377-85-5 | MFCD08868156 | 99.1% | 395.52 g/mol | C22H25N3O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1110985MG
Encompass_Preferred
GSK1702934A is a small-molecule activator selective for TRPC3 and TRPC6 channels used as a research tool to probe TRPC-mediated calcium signaling. It induces TRPC3/6-dependent currents and has been applied to studies of cardiac contractility and arrhythmogenesis in cellular and tissue models. The compound is offered in solid form and as DMSO solution for experimental convenience.
  • Selective TRPC3/6 activation for targeted functional studies.
  • High purity suitable for biochemical and electrophysiology assays.
  • Available as small solid quantities and as DMSO solution for convenience.
  • Supports studies of cardiac contractility and arrhythmogenesis.
  • Compatible with cellular assays and patch-clamp electrophysiology.
Catalog No. 50-002-64561