Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(1-Aminoethyl)phenol | 63720-38-7 | 98.0% | C8H11NO | 100 MG
SDP

Supplier:  Medchemexpress LLC HY70031100MG

Encompass_Preferred

3-(1-Aminoethyl)phenol is a chemical compound with the molecular formula C8H11NO, often used as a drug intermediate. It boasts a high purity of 98.0%. This compound is suitable for various laboratory applications.

  • High purity of 98.0%
  • Molecular formula: C8H11NO
  • CAS Number: 63720-38-7
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-34305


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.