Learn More
Medchemexpress LLC 3-(1-hydroxy-2-((4-phenylphenyl)methylamino)ethyl)phenol | 775294-71-8 | 99.8% | 5 MG

Supplier: Medchemexpress LLC HY1222145MG
AC-73 is a small-molecule inhibitor of cluster of differentiation 147 (CD147) used in biochemical and cellular research to disrupt CD147 dimerization. It is supplied as a white to off-white solid with high reported purity and good solubility in DMSO.
- High reported purity (99.84%).
- Molecular formula C21H21NO2 and molecular weight 319.4 g/mol.
- Soluble in DMSO at ≥ 250 mg/mL.
- Available in small research quantities and as a 10 mM DMSO solution.
- Recommended storage: powder at -20°C for long term; in solvent at -80°C for long term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.