Learn More
Medchemexpress LLC 3-[(1S)-1-(dimethylamino)ethyl]phenol | 139306-10-8 | 99.0% | 165.23 g·mol⁻¹ | C10H15NO | 10 G
SDP
Supplier:  Medchemexpress LLC HYW02057610G
Encompass_Preferred
Rivastigmine metabolite (NAP 226-90) is the primary human metabolite of rivastigmine and functions as an acetylcholinesterase (AChE) inhibitor. It is provided as a solid analytical standard with high reported purity and is intended for research use in analytical, pharmacological, and Alzheimer's disease studies.
  • Acts as an acetylcholinesterase inhibitor.
  • Supplied as a solid, white to off-white standard.
  • High reported purity suitable for analytical work.
  • Available in multiple pack sizes, including solid and solution formats.
  • Suitable as an analytical standard for LC-MS and quantitative assays.
  • Intended for use in pharmacology and Alzheimer's disease research.
Catalog No. 50-003-00739