Learn More
Medchemexpress LLC 3-(2-(2-aminoethoxy)ethoxy)propanoic acid | 791028-27-8 | MFCD20926377 | 99.7% | 177.20 | C7H15NO4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW040168500MG
Encompass_Preferred
Amino-PEG2-C2-acid is a cleavable three-unit polyethylene glycol (PEG) linker intended for research use in the synthesis of antibody-drug conjugates (ADCs) and PROTACs. It provides an amino-functionalized PEG spacer with a terminal propanoic acid group that facilitates conjugation chemistry and linker optimization in bioconjugation workflows.
  • Cleavable three-unit PEG spacer suitable for ADC and PROTAC synthesis.
  • Amino-functionalized for straightforward conjugation chemistry.
  • Terminal carboxylic acid enables diverse coupling strategies.
  • Reported high purity supports research applications.
  • Available in multiple package sizes, including a 500 mg option.
Catalog No. 50-002-03116