Learn More
Medchemexpress LLC 3-(2-((2-fluorophenyl)amino)oxazol-5-yl)benzoic acid | 2375281-44-8 | 99.8% | C16H11FN2O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13447710MG
Encompass_Preferred
NF-κB activator 2 is a small-molecule research compound that activates the NF-κB signaling pathway. It is provided as a dry powder or as a DMSO solution for biochemical and cell-based studies. Key physicochemical properties include molecular formula C16H11FN2O3, molecular weight 298.27, and high reported purity; follow recommended storage conditions for stability.
Catalog No. 50-002-60014