Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-[2,4-diamino-7-(3-hydroxyphenyl)pteridin-6-yl]phenol | 677297-51-7 | 99.4% | 346.34 g/mol | C18H14N6O2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY1011110MG

Encompass_Preferred

TG100-115 is a selective small-molecule inhibitor of PI3Kγ and PI3Kδ with nanomolar potency (IC50 83 nM and 235 nM, respectively). It is used in biochemical, cellular, and in vivo studies to investigate PI3Kγ/δ-mediated signaling, inflammatory responses, and cardioprotective effects.

  • Selective inhibition of PI3Kγ and PI3Kδ.
  • Nanomolar potency: PI3Kγ 83 nM, PI3Kδ 235 nM.
  • Limited activity against PI3Kα and PI3Kβ (>1 μM).
  • High purity (99.41%).
  • Molecular weight 346.34 g/mol; formula C18H14N6O2.
  • Soluble in DMSO (33.33 mg/mL) with warming and pH adjustment.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-13113


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.