Learn More
Medchemexpress LLC 3-(2,4-dimethoxyphenyl)-4-(3-thienyl)-1H-pyrrolo[2,3-b]pyridine | 1613710-01-2 | 99.7% | 200 MG

Supplier: Medchemexpress LLC HY120856200MG
ARN-3236 (CAS 1613710-01-2) is an orally active, selective inhibitor of salt-inducible kinase 2 (SIK2) used for research applications. It potently inhibits SIK2 at sub-nanomolar concentrations, demonstrates anti-cancer activity in cell-based assays, and can sensitize cancer cells to paclitaxel-class agents.
- Oral active selective SIK2 inhibitor.
- IC50 for SIK2 <1 nM; SIK1 21.63 nM; SIK3 6.63 nM.
- Purity greater than 98.0% (HPLC).
- Molecular weight 336.41 g/mol; formula C19H16N2O2S.
- Available as solid and DMSO solution; multiple sizes including 200 mg.
- Intended for research use only; not for human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.