Learn More
Medchemexpress LLC 3-[2-(4-fluorophenyl)ethynyl]-N-[3-(1H-pyrazol-4-yl)propyl]-4-(4-pyridinyl)-benzamide | 1800487-55-1 | ≥98.0% | 424.5 g/mol | C26H21FN4O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1362415MG
Encompass_Preferred
OT-82 is a small-molecule nicotinamide phosphoribosyltransferase (NAMPT) inhibitor used in preclinical research to deplete cellular NAD+ and selectively induce cell death in hematopoietic-derived malignancies. The compound is supplied as a solid for laboratory use and has been characterized in cell-free and cellular assays demonstrating potent activity against NAMPT.
  • Potent, selective NAMPT inhibitor suitable for research applications.
  • Demonstrated cytotoxicity in hematologic cancer cell lines with low-nanomolar IC50s.
  • Supplied as a solid, with reported purity of ≥98.0%.
  • Molecular weight 424.5 g/mol; molecular formula C26H21FN4O.
  • Intended for in vitro and preclinical studies; handle per SDS guidance.
Catalog No. 50-002-23246