Learn More
Medchemexpress LLC 3-[(2,6-dimethylphenoxy)methyl]-4-methoxybenzaldehyde | 438530-82-6 | 97.7% | 270.33 | C17H18O3 | 50 MG

Supplier: Medchemexpress LLC HYW15771950MG
3-[(2,6-dimethylphenoxy)methyl]-4-methoxybenzaldehyde is a synthetic chemical intermediate used in medicinal chemistry for the preparation of active pharmaceutical candidates. It is characterized by formula C17H18O3 and molecular weight 270.33 g/mol, and is supplied with quality documentation for research applications.
- Chemical intermediate for synthesis of active compounds.
- Molecular formula C17H18O3.
- Molecular weight 270.33 g/mol.
- High purity - 97.7% (as indicated on COA).
- Available in small-scale quantities suitable for R&D (50 mg to 10 g).
- Documentation includes certificate of analysis and handling information.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.