Learn More
Medchemexpress LLC 3-(2-chloro-6-methylphenyl)-7-((2-methoxy-4-(1-methylpiperidin-4-yl)phenyl)amino)-1-(5-methoxypyridin-2-yl)-... | 1936529-65-5 | 99.7% | 600.11 g/mol | C32H34ClN7O3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1011471MG
Encompass_Preferred
YKL-05-099 is a small-molecule inhibitor of salt-inducible kinases used in preclinical research. It shows high potency against SIK1 and SIK3 and moderate potency against SIK2, and is supplied in formats convenient for biochemical and cellular assays.
  • Potent inhibition of SIK1 and SIK3 (IC50 ≈ 10 nM and 30 nM).
  • Moderate inhibition of SIK2 (IC50 ≈ 40 nM).
  • High purity suitable for research applications.
  • Available as solid samples across multiple sizes and as a 10 mM solution in DMSO.
  • Molecular weight 600.11 g/mol and formula C32H34ClN7O3.
  • Suitable for biochemical and cellular assay use.
Catalog No. 50-002-11487