Learn More
Medchemexpress LLC 3-(2-Pyridyldithio)propanoic Acid | 68617-64-1 | 99.5% | 5 G
SDP
Supplier:  Medchemexpress LLC HY1301575G
Encompass_Preferred
3-(2-Pyridyldithio)propanoic Acid is an alkyl chain-based PROTAC linker designed for the synthesis of PROTACs.
  • Alkyl chain-based linker.
  • Connects two ligands: one for E3 ubiquitin ligase and one for a target protein.
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-40059