Learn More
Medchemexpress LLC 3-(2-((tert-butoxycarbonyl)amino)ethoxy)propanoic acid | 1260092-44-1 | MFCD22574796 | 97.0% | 233.26 | C10H19NO5 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1207751G
Encompass_Preferred
Boc-NH-PEG1-CH2CH2COOH is a Boc-protected, single-unit polyethylene glycol (PEG1) linker bearing a terminal carboxylic acid. It is intended for use in the synthesis of antibody-drug conjugates (ADCs) and proteolysis-targeting chimeras (PROTACs), providing a hydrophilic spacer and a protected amine for orthogonal chemistry. The material is supplied as a white to off-white solid with specified molecular weight and purity appropriate for research and linker construction.
  • Cleavable PEG1 spacer increases solubility and reduces steric hindrance.
  • Boc-protected amine enables orthogonal deprotection and coupling strategies.
  • Terminal carboxylic acid allows straightforward amide bond formation.
  • Compact linker length helps preserve biological activity of conjugates.
  • Packaged quantities suitable for lab-scale synthesis and conjugation workflows.
Catalog No. 50-002-01877