Learn More
Medchemexpress LLC 3,29-Dibenzoyl rarounitriol | 873001-54-8 | 666.93 | 5 MG
Supplier: Medchemexpress LLC HYN03575MG
3,29-Dibenzoyl rarounitriol is a major bioactive compound of multiflorane triterpene esters found in Trichosanthes kirilowii. It can be used as a marker for the quantitation of Trichosanthes kirilowii.
- High purity of 98.96%.
- Documentation available, including data sheet, SDS, and COA.
- Soluble in DMSO and ethanol for in vitro applications.
- Stable under sealed storage conditions away from moisture and light.
- Suitable for research use only.