Learn More
Medchemexpress LLC 3(2H)-Isothiazolone, 2-(1,1-dimethylethyl)-5-phenyl-4-[[4-(1-piperidinyl)phenyl)amino]-, 1,1-dioxide | 898800-26-5 | 99.77% | 439.57 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1828210MM/1ML
Encompass_Preferred
AZ876 is a selective, orally active agonist of liver X receptor (LXRα/β) that regulates lipid metabolism and anti-inflammatory effects. It induces the expression of target genes such as ABCA1 and ABCG1, and promotes reverse cholesterol transport (RCT). This product increases cardiac polyunsaturated fatty acid levels, reduces myocardial fibrosis, and decreases lesion area and monocyte adhesion in atherosclerosis models.
  • Useful in cardiovascular disease research
  • Prevents and treats β-adrenergic-induced cardiac diastolic dysfunction
  • Inhibits atherosclerosis progression
  • Time-dependently induces LXR target genes in HL-1 cardiomyocytes
Catalog No. 50-003-87604