Learn More
Medchemexpress LLC 3(2H)-Isothiazolone, 5-chloro-2-methyl-, mixt. with 2-methyl-3(2H)-isothiazolone | 55965-84-9 | MFCD01716911 | 14.5% | 264.75 | C8H9ClN2O2S2 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW017982500G
Encompass_Preferred
Kathon 886MW (14.5% in water) is a ready-to-use aqueous biocide containing a CMIT/MIT isothiazolinone mixture. It provides broad-spectrum antibacterial and antifungal control for preservation and microbial management in water-based systems such as metalworking fluids. Supplied for research and industrial applications; not for human therapeutic use.
  • Broad-spectrum antibacterial and antifungal activity.
  • Ready-to-use 14.5% aqueous formulation.
  • Suitable for metalworking fluids and other water-based systems.
  • Helps prevent microbial contamination and product spoilage.
  • Provided with datasheet and SDS for safe handling and use.
Catalog No. 50-003-01599