Learn More
AVANTOR MASTERFLEX (VWR LLC) Catalyst Stopped Tubing Assembly, Platinum-Cured Silicone, 3.2 mm ID, 6 pk
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9710016
Encompass
Start pumping with little effort, as preconfigured links aid quick and easy setup. Ensure the optimal amount of tubing is in the pump head. Use with the Catalyst extension coils / tubing to complete your system.
Catalog No. NC1185960