Learn More
Medchemexpress LLC 3,3'-((2-amino-2-((2-carboxyethoxy)methyl)propane-1,3-diyl)bis(oxy))dipropanoic acid | 174362-95-9 | MFCD31615450 | 98.0% | 337.32 | C13H23NO9 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1175195G
Encompass_Preferred
Amino-Tri-(carboxyethoxymethyl)-methane (CAS 174362-95-9) is a small-molecule reagent used as an ADC linker intermediate and in peptide and bioconjugation synthesis. It has a reported purity of 98.0%, molecular formula C13H23NO9, and molecular weight 337.32 g/mol. Recommended storage: 4°C, protect from light; in solvent: -80°C (6 months), -20°C (1 month).
  • Small-molecule ADC linker intermediate for bioconjugation
  • High reported purity suitable for synthetic applications
  • Available in multiple laboratory package sizes including 5 g
  • Stable under low-temperature storage when protected from light
  • Applicable to peptide synthesis and linker development workflows
Catalog No. 50-002-28726