Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 3,3,3-Trifluoropropane-1-sulfonyl chloride, 845866-80-0, MFCD06248898, 1g

Supplier:  Matrix Scientific 0126161G

Encompass_Preferred

Molecular Formula, C3H4ClF3O2S, Purity 95%, Molecular Weight 196.58, Boiling Point 76-78/16mm

Catalog No. 50-012-5965


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.