Learn More
Medchemexpress LLC 3-(3,4-dichlorophenyl)-1-methoxy-1-methylurea | 330-55-2 | MFCD00055249 | 99.9% | 249.09 g/mol | C9H10Cl2N2O2 | 10 MG

Supplier: Medchemexpress LLC HYB186610MG
Linuron is a phenylurea herbicide and photosystem II inhibitor supplied as a high-purity solid for laboratory and analytical research. It is intended for use as a reference standard in biochemical and environmental studies where a well-characterized herbicide is required.
- High purity: 99.9%
- Molecular weight: 249.09 g/mol
- Chemical formula: C9H10Cl2N2O2
- Appearance: white to off-white solid
- Available in small quantities suitable for analytical use (e.g., 10 mg)
- Includes datasheet, certificate of analysis, and safety data sheet
- Stable when stored as powder at -20°C or 4°C per recommended conditions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.