Learn More
Medchemexpress LLC 3-(3,4-dimethoxyphenyl)propanoic acid | 2107-70-2 | MFCD00002774 | 5g
SDP
Supplier:  Medchemexpress LLC HYY16205G
Encompass_Preferred
3-(3 4-Dimethoxyphenyl)propanoic acid is an orally active short-chain fatty acids (SCFAs) 3-(3 4-Dimethoxyphenyl)propanoic acid stimulates globin gene expression erythropoiesis in vivo and is used for the hemoglobinopathies and other anemias[1]
Catalog No. 50-002-67843