Learn More
Cayman Chemical 3 3 5-TrIIodo-L-thyronIn 1g
SDP
Supplier:  Cayman Chemical 309961 G
Encompass_Preferred
A thyroid hormone; agonist of TRα and TRβ (Kis = 2.3 nM for both); modulates diverse biological properties, including metabolism, cardiac function, growth, and redox signaling; inhibits LAT1-mediate leucine uptake in HEK293 cells at 10 µM
Catalog No. 50-298-9685