Learn More
Medchemexpress LLC 3-[[(3-Amino-3-carboxypropyl)methoxyphosphinyl]oxy]benzeneacetic acid | 926281-37-0 | ≥95% | 331.26 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY108467
Encompass_Preferred
GGsTop (Nahlsgen) is a potent, non-toxic, highly selective, and irreversible γ-glutamyl transpeptidase (GGT) inhibitor. It shows a Ki of 170 μM for Human GGT and exhibits Kons of 150 M-1 s-1 against E.coli GGT and 51 M-1 s-1 against human GGT. GGsTop protects against hepatic ischemia-reperfusion injury in a rat model. It does not inhibit glutamine amidotransferases or asparagine synthetase, and suppresses oxidative stress by inhibiting GGT activation. It also prevents ischemia/reperfusion-induced acute kidney injury in rats in vivo.
  • Potent, selective, and irreversible γ-glutamyl transpeptidase (GGT) inhibitor
  • Non-toxic compound
  • Protects against hepatic ischemia-reperfusion injury
  • Suppresses oxidative stress by inhibiting GGT activation
  • Prevents ischemia/reperfusion-induced acute kidney injury
Catalog No. 50-003-47292