Learn More
Medchemexpress LLC 3,3′-bisdemethylpinoresinol | 340167-81-9 | 99.0% | 330.33 g/mol | C18H18O6 | 5mg
SDP
Supplier:  Medchemexpress LLC HYW7451645MG
Encompass_Preferred
3 3 -Bisdemethylpinoresinol lignin is a nature product and has MMP-1 inhibitory activity in UVA-irradiated human dermal fibroblasts 3 3 -Bisdemethylpinoresinol can be isolated from the seeds of Morinda citrifolia[1]
Catalog No. 50-002-69614