Learn More
Medchemexpress LLC 3-(3-chloro-phenoxymethyl)-1-(tetrahydro-pyran-4-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-ylamine | 1188296-52-7 | MFCD22123245 | 99.1% | 359.81 g·mol⁻¹ | C17H18ClN5O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1247010MG
Encompass_Preferred
PF-4800567 is a selective small-molecule inhibitor of casein kinase 1 epsilon (CK1ε) used in research to probe CK1 signaling and circadian rhythm pathways. It potently inhibits CK1ε (IC50 = 32 nM) with greater than 20-fold selectivity over CK1δ and is supplied as research-grade material for biochemical and cell-based assays.
  • Potent CK1ε inhibition (IC50 32 nM).
  • Greater than 20-fold selectivity over CK1δ (IC50 711 nM).
  • Molecular weight 359.81 g·mol⁻¹ and formula C17H18ClN5O2.
  • High reported purity (~99.1%).
  • Suitable for biochemical and cell-based studies of CK1 signaling and circadian biology.
Catalog No. 50-002-36358