Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3-Diaminobenzidine Tetrahydrochloride, MP Biomedicals™

3,3-Diaminobenzidine Tetrahydrochloride is a reagent used in colorimetric analysis of Se and other metals.

$366.00 - $660.00

Specifications

Form Powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
02-150-82505
View Documents
MP Biomedicals, Inc
0215082505
5 g
Each for $366.00
Add to Cart
 
02-150-82510
View Documents
MP Biomedicals, Inc
0215082510
10 g
Each for $660.00
Add to Cart
 
Description

Description

  • 3,3-Diaminobenzidine Tetrahydrochloride is a reagent used in colorimetric analysis of Se and other metals.
  • DAB is utilized in many applications for visualization of peroxidase activity.
  • In the peroxidase reaction, DAB serves as a hydrogen donor in the presence of peroxide; the oxidized DAB forms an insoluble brown end-product.
Specifications

Specifications

Powder
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.