Learn More
3,3-Diaminobenzidine Tetrahydrochloride, MP Biomedicals™
3,3-Diaminobenzidine Tetrahydrochloride is a reagent used in colorimetric analysis of Se and other metals.
Supplier:  MP Biomedicals, Inc 0215082505
Quantity 5 g
 View more versions of this product
Catalog No. 02-150-82505
Add to Cart
Description
Description
  • 3,3-Diaminobenzidine Tetrahydrochloride is a reagent used in colorimetric analysis of Se and other metals.
  • DAB is utilized in many applications for visualization of peroxidase activity.
  • In the peroxidase reaction, DAB serves as a hydrogen donor in the presence of peroxide; the oxidized DAB forms an insoluble brown end-product.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Powder
5 g
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov