Learn More
AdipoGen 3,3'-Diethylthiacarbocyanine iodide (250 mg)
SDP
Supplier:  AdipoGen CDXD0447M250
Encompass_Preferred
Chemical. CAS: 905-97-5. Formula: C21H21IN2S2. MW: 492.44. Synthetic. DiSC2(3) is a light-sensitive cyanine dye. Spectral data: Absorption: lambdamax 560nm (MeOH). Cyanine dyes are commonly used for photographic sensitisation, dye lasers and fluorescence microscopy. A different salt form of Thiazole purple (bromide salt). DiSC2(3) has been shown to bind to PNA-DNA complex and accelerated photobleaching can be used for diagnostics purpose. It has been used as an acceptor dye in electronic excitation energy transfer (EEET).
Catalog No. 50-205-4390