Learn More
Medchemexpress LLC 3,3'-Difluorobenzaldazine | 15332-10-2 | MFCD03653615 | 99.4% | 244.24 | C14H10F2N2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY146115MG
Encompass_Preferred
3,3'-Difluorobenzaldazine (DFB) is a small-molecule research compound reported as a selective positive allosteric modulator of the metabotropic glutamate receptor 5 (mGluR5). It is intended for preclinical and in vitro research and is characterized by defined molecular properties and high batch purity.
  • Selective positive allosteric modulator of mGluR5.
  • High purity suitable for research applications.
  • Available in multiple small-scale pack sizes for screening and dose-response studies.
  • Molecular formula C14H10F2N2; molecular weight 244.24.
  • Supplied with batch-specific certificate of analysis for traceability.
Catalog No. 50-002-22741