Learn More
Cayman Chemical 3 3-DIhexyloxacarbocynIn 500mg
SDP
Supplier:  Cayman Chemical 23124500 MG
Encompass_Preferred
A fluorescent dye; ex/em spectra = 484/501 nm, respectively; accumulates in ER at high concentrations and has been used to visualize ER in moss, yeast, and muscle cells; accumulates in mitochondria at low concntrations and has been used to assess mitochondrial dislocations, fusion, and fission in living cells, as well as to visualize cellular apoptosis; used to assess cell membrane potential, as depolarization directly correlates with increased fluorescence; photoexcitation inactivates intracellular organelle movement and inhibits microtubule polymerization in vitro
Catalog No. 50-299-4054