Learn More
Cayman Chemical 3 3-DIIndolylmethn 100mg
SDP
Supplier:  Cayman Chemical 15927100 MG
Encompass_Preferred
Phytochemical from cruciferous vegetables that demonstrates anticancer and chemopreventative effects (10-30 µM) involving the induction of Phase 2 enzymes, promotion of apoptosis, induction of cell cycle arrest, inhibition of cell proliferation, and inhibition of histone deacetylases and DNA methylation activities
Catalog No. 50-297-1041