Learn More
Medchemexpress LLC 3,3-Dimethylacrylic acid | 541-47-9 | 99.99% | 100.12 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01061150G
Encompass_Preferred
3,3-Dimethylacrylic acid is an active small molecule that can be used as a drug intermediate. It exhibits antituberculosis activity with a MIC of 0.078 μg/mL and can be used for the synthesis of triorganotin(IV) complexes of cinnamic acid. It is for research use only and not sold to patients. It has an appearance of a solid, white to off-white color. It is classified under Ketones, Aldehydes, Acids and its initial source is an endogenous metabolite.
  • Exhibits antituberculosis activity with a MIC of 0.078 μg/mL.
  • Can be used for the synthesis of triorganotin(IV) complexes of cinnamic acid.
  • For research use only.
  • Appearance: solid, white to off-white color.
  • Classified under ketones, aldehydes, acids.
  • Initial source is an endogenous metabolite.
Catalog No. 50-004-51896