Learn More
Ambeed 3,3-Dimethylmorpholine | 59229-63-9 | MFCD09608072 | 250mg
SDP
Supplier:  Ambeed A334648250MG
Encompass_Preferred
3,3-Dimethylmorpholine | Putity: 97% | MW: 115.17 | 59229-63-9 | MFCD09608072 | 250mg
Catalog No. NC1764180