Learn More
Medchemexpress LLC HY-D0969 5mg Medchemexpress, 3,3'-Dioctadecyloxacarbocyanine perchlorate CAS:34215-57-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD09695MG
Encompass_Preferred
Medchemexpress, HY-D0969 5mg 3,3'-Dioctadecyloxacarbocyanine perchlorate CAS:34215-57-1 3,3′-Dioctadecyloxacarbocyanine perchlorate is a green fluorescent lipophilic tracer, which is weakly fluorescent in water but highly fluorescent and quite photostable when incorporated into membranes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3216