Learn More
Cayman Chemical 3 3-Dioctadecyloxacarbocyani
SDP
Supplier:  Cayman Chemical 2787650 MG
Encompass_Preferred
A lipophilic fluorescent dye; ex/em = 484/501 nm, respectively; has been used in the clonal analysis of S. purparatus embryos
Catalog No. 50-293-7263