Learn More
Medchemexpress LLC 3,3'-Dipropylthiadicarbocyanine iodide | 53213-94-8 | MFCD00075033 | 99.4% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYD0085100MG
Encompass_Preferred
DiSC3(5) is a fluorescent probe commonly used as a tracer dye to evaluate mitochondrial membrane potential. It inhibits the respiratory system associated with mitochondrial NAD, and can induce membrane hyperpolarization of Ehrlich ascites tumor cells in the presence of Na+/K+-ATPase inhibitor ouabain 2.
  • Fluorescent probe
  • Tracer dye to evaluate mitochondrial membrane potential
  • Excitation/emission wavelength up to 622/670 nm
  • Inhibits the respiratory system associated with mitochondrial NAD (IC50 value is 8 μM)
  • Induces membrane hyperpolarization of Ehrlich ascites tumor cells in the presence of Na+/K+-ATPase inhibitor ouabain 2
Catalog No. 50-004-10811