Learn More
Cayman Chemical 3 3-SulfInyldIpropnoIc AcId 1g
SDP
Supplier:  Cayman Chemical 372141 G
Encompass_Preferred
An APE1/Ref-1 inhibitor (IC50 = 40 µM); a building block in the synthesis of DSSO
Catalog No. 50-291-5376