Learn More
Medchemexpress LLC 3-(3-Trifluoromethylphenyl)propanol | 78573-45-2 | 99.98% | C10H11F3O | 1 G
SDP
Supplier:  Medchemexpress LLC HY760391G
Encompass_Preferred
3-(3-Trifluoromethylphenyl)propanol is a chemical compound primarily used as a drug intermediate for the synthesis of various active compounds. This product is intended for research use only and has not been fully validated for medical applications.
  • Used in laboratory chemical manufacturing
  • Recommended storage at 4°C, sealed, away from moisture
  • In solvent, stable at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-34131