Learn More
Medchemexpress LLC 3-(3-Trifluoromethylphenyl)propanol | 78573-45-2 | C10H11F3O | 100 G
SDP
Supplier:  Medchemexpress LLC HY76039100G
Encompass_Preferred
3-(3-Trifluoromethylphenyl)propanol is a drug intermediate used for the synthesis of various active compounds. It is stable under recommended storage conditions and has a molecular weight of 204.19.
  • Used as a drug intermediate
  • Stable under recommended storage conditions
  • Suitable for laboratory chemicals and manufacture of substances
  • For research use only
Catalog No. 50-004-34169